Anti-GPAM Rabbit Polyclonal Antibody
ABGEAP10150C
: Abgent
- Antibody type:Primary
- Antigen name:Glycerol-3-phosphate acyltransferase, mitochondrial
- Antigen symbol:GPAM
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-Terminal
- Cross adsorption:No
- Antigen synonyms:Glycerol-3-phosphate acyltransferase 1|KIAA1560|mitochondrial|GPAT1|GPAT-1|GPAT
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Type: Primary
Antigen: GPAM (glycerol-3-phosphate acyltransferase, mitochondrial)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human