Anti-COX7A2 Rabbit Polyclonal Antibody
AVIVARP66548_P050
- Antibody type:Primary
- Antigen name:cytochrome c oxidase subunit VIIa polypeptide 2 (liver)
- Antigen symbol:COX7A2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:1347
- Antigen synonyms:COXVIIAL|COX7AL|COX7AL1|COXVIIa-L|VIIAL
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: COX7A2 (cytochrome c oxidase subunit VIIa polypeptide 2 (liver))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human