Anti-EBNA1BP2 Rabbit Polyclonal Antibody
Catalog # AVIVARP64762_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:EBNA1 binding protein 2
- Antigen symbol:EBNA1BP2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:10969
- Antigen synonyms:EBP2|NOBP|P40
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: EBNA1BP2 (EBNA1 binding protein 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human