Anti-PPP1CC Rabbit Polyclonal Antibody
AVIVARP64668_P050
- Antibody type:Primary
- Antigen name:Protein phosphatase 1, catalytic subunit, gamma
- Antigen symbol:PPP1CC
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:5501
- Antigen synonyms:PP1C|PP-1G|PPP1G
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: PPP1CC (protein phosphatase 1, catalytic subunit, gamma isozyme)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human