Anti-CAP2 Rabbit Polyclonal Antibody
AVIVARP63855_P050
- Antibody type:Primary
- Antigen name:CAP, adenylate cyclase-associated protein, 2 (yeast)
- Antigen symbol:CAP2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:10486
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: CAP2 (CAP, adenylate cyclase-associated protein, 2 (yeast))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: