Anti-GABBR2 Rabbit Polyclonal Antibody
Catalog # AVIVARP61115_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:gamma-aminobutyric acid (GABA) B receptor, 2
- Antigen symbol:GABBR2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:9568
- Antigen synonyms:GABABR2|HRIHFB2099|GPRC3B|GPR51|HG20
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: GABBR2 (gamma-aminobutyric acid (GABA) B receptor, 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: