Anti-AP2A2 Rabbit Polyclonal Antibody
Catalog # AVIVARP59974_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:adaptor-related protein complex 2, alpha 2 subunit
- Antigen symbol:AP2A2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human,Guinea pig,Dog,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:161
- Antigen synonyms:HYPJ|HIP-9|ADTAB|HIP9|CLAPA2
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: AP2A2 (adaptor-related protein complex 2, alpha 2 subunit)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Dog, Guinea pig, Human, Mouse, Rat