Anti-ZER1 Rabbit Polyclonal Antibody
AVIVARP59408_P050
- Antibody type:Primary
- Antigen name:zyg-11 related, cell cycle regulator
- Antigen symbol:ZER1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:middle
- Cross adsorption:none
- Gene ID:10444
- Antigen synonyms:ZYG11BL|C9orf60|ZYG
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: ZER1 (zyg-11 related, cell cycle regulator)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: