Anti-ZNF134 Rabbit Polyclonal Antibody
AVIVARP58103_P050
- Antibody type:Primary
- Antigen name:zinc finger protein 134
- Antigen symbol:ZNF134
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:middle
- Cross adsorption:none
- Gene ID:7693
- Antigen synonyms:pHZ-15
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: ZNF134 (zinc finger protein 134)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human