Anti-ADRBK1 Rabbit Polyclonal Antibody
AVIVARP51966_P050
- Antibody type:Primary
- Antigen name:adrenergic, beta, receptor kinase 1
- Antigen symbol:ADRBK1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Pig,Human,Bovine,Guinea pig,Dog,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:156
- Antigen synonyms:BARK1|BETA-ARK1|GRK2
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: ADRBK1 (adrenergic, beta, receptor kinase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Bovine, Dog, Guinea pig, Human, Mouse, Pig, Rat