Order Entry
Austria
ContactUsLinkComponent
Anti-AP1G1 Rabbit Polyclonal Antibody
Anti-AP1G1 Rabbit Polyclonal Antibody
Catalog # AVIVARP51923_P050
Anti-AP1G1 Rabbit Polyclonal Antibody
Catalog # AVIVARP51923_P050

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    adaptor-related protein complex 1, gamma 1 subunit
  • Antigen symbol:
    AP1G1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
  • Reactivity:
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Epitope:
    C-terminal
  • Cross adsorption:
    none
  • Gene ID:
    164
  • Antigen synonyms:
    CLAPG1|ADTG
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    50 µG

Specifications

About this item

Type: Primary
Antigen: AP1G1 (adaptor-related protein complex 1, gamma 1 subunit)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: