Anti-NR2E1 Rabbit Polyclonal Antibody
Catalog # AVIVARP45613_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:nuclear receptor subfamily 2, group E, member 1
- Antigen symbol:NR2E1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:7101
- Antigen synonyms:TLL|XTLL|TLX
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: NR2E1 (nuclear receptor subfamily 2, group E, member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human