Anti-LRPAP1 Rabbit Polyclonal Antibody
AVIVARP45276_T100
- Antibody type:Primary
- Antigen name:Low density lipoprotein receptor-related protein associated protein 1
- Antigen symbol:LRPAP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:4043
- Antigen synonyms:MRAP|HBP44|A2MRAP|A2RAP|RAP|MYP23|alpha-2-MRAP
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Type: Primary
Antigen: LRPAP1 (low density lipoprotein receptor-related protein associated protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: