Order Entry
Austria
ContactUsLinkComponent
 
Anti-AWAT1 Rabbit Polyclonal Antibody
  AVIVARP44651_P050
undefined
Anti-AWAT1 Rabbit Polyclonal Antibody
  AVIVARP44651_P050

 

  • Antibody type:
    Primary
  • Antigen name:
    acyl-CoA wax alcohol acyltransferase 1
  • Antigen symbol:
    AWAT1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
  • Reactivity:
    Human,
    Bovine,
    Guinea pig,
    Dog,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Epitope:
    N-terminal
  • Cross adsorption:
    none
  • Gene ID:
    158833
  • Antigen synonyms:
    DGAT2L3|DGA2
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    50 µG

 

 

Type: Primary
Antigen: AWAT1 (acyl-CoA wax alcohol acyltransferase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Bovine, Dog, Guinea pig, Human, Mouse, Rat