Anti-RAE1 Rabbit Polyclonal Antibody
Catalog # AVIVARP40301_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Ribonucleic Acid Export 1
- Antigen symbol:RAE1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:8480
- Antigen synonyms:MIG14|dJ481F12.3|MRNP41|Mnrp41|dJ800J21.1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: RAE1 (ribonucleic acid export 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: