Anti-SP140L Rabbit Polyclonal Antibody
AVIVARP36135_T100
- Antibody type:Primary
- Antigen name:SP140 nuclear body protein-like
- Antigen symbol:SP140L
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:middle
- Cross adsorption:none
- Gene ID:93349
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Type: Primary
Antigen: SP140L (SP140 nuclear body protein-like)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human