Anti-ZBTB18 Rabbit Polyclonal Antibody
Catalog # AVIVARP35799_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:zinc finger and BTB domain containing 18
- Antigen symbol:ZBTB18
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:10472
- Antigen synonyms:ZNF238|C2H2-171|MRD22|RP58|TAZ-1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: ZBTB18 (zinc finger and BTB domain containing 18)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: