Safety cabinets for chemicals, CS-Classic
: asecos
Safety storage cabinets for non-flammable chemicals and toxic substances in workrooms.
- Body made of sheet steel with high grade structured paint finish with robust construction and longevity
- Lockable doors with cylinder locking integrated in the turning handle
- Adusting aids to compensate for uneven floor
- Natural ventilation due to vent openings at the cabinet bottom for the reduction of odours
: Supplied without interior equipment packages. Interior equipment packages need to be ordered in addition. Please check the website or contact Avantor sales office for further information.
Frequently Bought Together