Pendulum hardness tester
Supplier: Industrial Physics
Certificates
About this item
Stainless steel.
- Easy menu driven interface with jog dial
- Design 'solid supports' to level the instrument
- Integrated storage of glass plate and pendulum
- Menu driven calibration menu eases both Persoz and König method
- The end of a test is indicated by a visual and acoustic signal
The pendulum hardness test is based on the principle that the amplitude of oscillation of a pendulum touching a surface decreases more rapidly the softer the surface. It has a wide range of applications areas including laboratories, coating industry, paint industry, and the automotive industry.
Certifications: ISO 1522 (2005) standard for the correct execution of the test.
Frequently Bought Together