Order Entry
Austria
ContactUsLinkComponent
Small cotton swab
Small cotton swab
Catalog # PMPC868-WCS
Supplier:  PURITAN MEDICAL
undefined
Small cotton swab
Catalog # PMPC868-WCS
Supplier:  PURITAN MEDICAL

Specifications

  • Handle length:
    148 mm
  • Head length:
    11 mm
  • Description:
    Small applicators
  • Environmentally Preferable:
  • Shaft/head:
    Wood/cotton
  • Pk:
    10.000

Specifications

About this item

The cotton-tipped applicator is produced with high quality USP grade cotton.

  • Bonded securely to a sturdy handle by an aqueous based adhesive
  • Small tip for hard-to-reach or small areas
  • Carded and coiled onsite to maintain consistency

Suitable for cleaning medical devices, to remove oil or dirt from machine parts, in veterinary care, and the cleaning of sensors or electronic components.

Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904