ContactUsLinkComponent
UPLC columns, ACQUITY UPLC™ BEH C8
UPLC columns, ACQUITY UPLC™ BEH C8
Supplier:  WATERS
Certificates
UPLC columns, ACQUITY UPLC™ BEH C8
Supplier:  WATERS
Specifications
  • Catalog No:
  • WATE186002877
  • WATE186002876
  • Carbon load:
  • 13%
  • 13%
  • Endcapped:
  • Yes, Yes
  • Yes, Yes
  • pH range:
  • 1 - 12
  • 1 - 12
  • Surface area:
  • 185 m²/g
  • 185 m²/g
  • USP code:
  • L7, L7
  • L7, L7