Viscosity flow cups
Supplier: Industrial Physics
Certificates
About this item
These viscosity flow cups are titanium anodised aluminum viscosity cups.
Suitable to be used with stand, to measure the viscosity of lacquers, paint and other liquids.
Certifications: AFNOR
Frequently Bought Together