4-Sulphobenzoic acid monopotassium salt 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 4-Sulphobenzoic acid potassium salt
CAS Number:5399-63-3
:
Formula:C₇H₅KO₅S
MDL Number:MFCD00007509
Molecular Weight:240.28 g/mol
EINECS:226-435-8
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together