Panoramic autoclavable goggles, BCAP series, BioClean Clearview™
111-2139
: Nitritex
- Type:Cleanroom, autoclavable, anti-scratch
- Colour:White
- Description:Autoclavable panoramic goggles
- Environmentally Preferable:
- Lenses:Clear PC
- Pk:12
BioClean Clearview™ autoclavable cleanroom goggles have a super-soft thermoplastic rubber frame to provide wearer comfort.
- Toughened anti-scratch, anti-fog lens
- Super-soft frame
- Indirect ventilation system
- Ideal for wearing over eyeglasses
- Non-linting, latex-free headband
- Tested to withstand 40 autoclave cycles of 30 minutes duration at 121 °C/250 °F (under laboratory conditions)
- Anti-fog performance remaining for up to 25 cycles with no degradation (under laboratory conditions)
BioClean Clearview™ Autoclavable Panoramic Goggles have a toughened, anti-scratch, anti-fog lens providing excellent optical clarity even after multiple autoclave cycles. The goggles feature a super-soft frame for comfort and an indirect ventilation system to reduce the risk of contamination entering the controlled environment. The extra-wide and deep lens offers the wearer increased field of vision and are ideal for wearing over large eyeglasses.
EN 166:2001 Optical Class 1; PPE Directive 89/686/EEC Cat II
: Manufactured in a facility holding ISO 9001:2015. Processed in an NEBB certified ISO Class 4 cleanroom environment. Conforms to Category 2 – Personal Protective Equipment PPE Regulation (EU)2016/425. Complies with requirements of EN 166:2001 standard for personal eye protection.Tested to ANSI/ISEA Z87.1-2010, the American National Standard for Occupational and Educational Personal Eye and Face Protection Devices (BCAP-1 Goggle Style Only).
: Goggle (BCA and BCAP-1): One piece per sealed PE bag (to be removed prior to autoclaving); 12 bags per inner box; five boxes per lined carton (60 pieces). Replacement lens (BCAP-SL): One piece per sealed PE bag; 10 bags per inner box; 10 boxes per lined carton (100 pieces). Replacement strap (BCAP-SS): 25 pieces per sealed PE bag; two bags per inner box; 10 boxes per lined carton (500 pieces).