asecos, safety storage cabinets, S-Classic
: asecos
Fireproof safety cabinets, classified type 90 (fire resistant up to 90 minutes).
- Robust construction and longevity - safety elements assembled outside the storage compartment for increased protection against corrosion, easy to clean
- Convenient handling: Doors can be opened with minimal effort
- User friendly and safe: Door open arrest system (optional), convenient and safe removal or storage of containers
- No unauthorised use: Doors lockable with cylinder locking (integration in a locking system possible)
- Easy alignment: Adjusting aids to compensate for uneven floor
- Quickly put into operation: Integrated air ducts ready for connection (NW 75 mm) to a technical exhaust system
Outer body made of sheet steel with high grade structured paint finish, interior with chemically resistant surface.
: Furnace tested (type test) in accordance with EN 14470-1
Fire resistance 90 minutes
GS approval, CE conformity
EN 14727 conformity (laboratory furniture)
Made in Germany
Fire resistance 90 minutes
GS approval, CE conformity
EN 14727 conformity (laboratory furniture)
Made in Germany
: Other colours are available on request. Supplied without interior equipment packages. Interior equipment packages need to be ordered separately. Please check the website or contact Avantor for further information.
: Delivery prices available upon request.
Frequently Bought Together