Pallets and sumps for IBC storage
: asecos
Blue polyethylene sump pallet with 1100 L capacity for the storage of IBC.
- Compact and robust design
- Also available with an integrated pallet shape storage platform made of polyethylene
- Ideal for storing 1000 litres IBC or small containers
Approved for the safe storage of water pollutants.
Ideal for storing liquids with a maximum specific weight of up to 1800 kg/m³.
Frequently Bought Together