Order Entry
Austria
ContactUsLinkComponent
 
Gel filtration columns, HiPrep™ with Sephacryl® S-200 HR
 :  Cytiva
 
undefined
Gel filtration columns, HiPrep™ with Sephacryl® S-200 HR
 :  Cytiva

 

  • Catalog No:
  • 17-1166-01
  • 17-1195-01
  • Max. pressure:
  • 1,5 bar
  • 1,5 bar
  • pH range:
  • 3 - 11
  • 3 - 11
  • Storage:
  • 4 to 30 °C, 20% Ethanol
  • 4 to 30 °C, 20% Ethanol

 

Frequently Bought Together