Order Entry
United Kingdom
ContactUsLinkComponent
High content cell imaging system, micro confocal, ImageXpress
High content cell imaging system, micro confocal, ImageXpress
Supplier:  Molecular Devices
Certificates
undefined
High content cell imaging system, micro confocal, ImageXpress
Supplier:  Molecular Devices

Specifications

  • Catalog No:
  • MLDV5041314
  • MLDV5041310
  • MLDV5041309
  • 735-0364
  • MLDV5041312
  • MLDV5040287
  • MLDV5039731
  • MLDVMICROCONFO_REF
  • Condenser:
  • 65 mm WD and PhL, Ph1, and Ph2 selectable phase rings
  • 65 mm WD and PhL, Ph1, and Ph2 selectable phase rings
  • Sensor:
  • High quantum efficiency 16 bit scientific CMOS
  • High quantum efficiency 16 bit scientific CMOS
  • Electrical:
  • 240 V
  • 240 V
  • W×D×H:
  • 530×860×480 mm
  • 530×860×480 mm
  • Weight:
  • 104 kg
  • 104 kg

Specifications

Objectives

Product Information

Frequently Bought Together