Eyepiece cameras, Dino-Eye
Supplier: Dino-Lite / IDCP B.V.
Certificates
About this item
With these cameras users can turn their conventional microscopes into digital microscopes.The ocular (eyepiece) of the non-digital microscope is simply replaced by an Dino-Eye eyepiece and connected to a computer via USB.
- Different models for different eyepiece sizes
- High speed USB 2.0 connection
- Software included for image or video processing, incl. calibrated measurement features
- MicroTouch feature (model AM4023CT only): Touch sensitive area that users can use to capture images instead of using a keyboard and mouse
- Model AM7025X: Edge series with improved colour accuracy and larger field of view
- Models AM4023CT and AM7025X also connect to devices with C-mount
Edge model is suitable for demanding applications where true colour reproduction is of the utmost importance, like with pathology samples or quality control.
Delivery information: Supplied with carrying pouch, software CD (DinoCapture (Windows) and DinoXcope (MAC)), calibration target and user manual.
Frequently Bought Together